Formaldehyde diethyl acetal ≥99.7%

Synonyms: Ethylal, Diethoxymethane

538280-4L 538280-500ML
220076-540EA 111.38 USD
220076-540 220076-541
Formaldehyde diethyl acetal ≥99.7%
Formaldehyde diethyl acetal
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 88 °C (1013 hPa)
Melting Pt: –66 °C
Density: 0.8319 g/cm³ (20 °C)
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00009246
CAS Number: 462-95-3
EINECS: 207-330-6
UN: 2373
ADR: 3,II

Order Now

Specification Test Results

Assay 99.7%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR