2,2-Dimethyl-1,3-propanediol ≥99%

Danger

Synonyms: Neopentyl Glycol, 2,2-Dimethylpropane-1,3-diol

538256-500G 538256-3KG 538256-25G
220076-533EA 36.4 USD
220076-533 220076-532 220076-531
2,2-Dimethyl-1,3-propanediol ≥99%
2,2-Dimethyl-1,3-propanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 128…130 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004685
CAS Number: 126-30-7
EINECS: 204-781-0
Merck Index: 12,06547

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR