Propionaldehyde ≥97%, Reagent Grade

Danger

Synonyms: Propanal

538124-250ML 538124-1L
220076-467EA 29.48 USD
220076-467 220075-586
Propionaldehyde ≥97%, Reagent Grade
Propionaldehyde
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48 °C (1013 hPa)
Melting Pt: –81 °C
Density: 0.811 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 13,07915

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR