2-(2-Ethoxyethoxy)ethyl acetate ≥99%

Synonyms: Diethylene glycol monoethyl ether acetate

537527-4L 537527-1L 537527-20L
220076-401EA 259.54 USD
220076-401 220076-399 220076-400
2-(2-Ethoxyethoxy)ethyl acetate ≥99%
2-(2-Ethoxyethoxy)ethyl acetate
Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1.0114 g/cm³ (20 °C)
MDL Number: MFCD00041928
CAS Number: 112-15-2
EINECS: 203-940-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR