Isopropyl acetate ≥99.6%

Danger

Synonyms: Acetic acid isopropyl ester

537462-4L 537462-1L
220075-093EA 85.85 USD
220075-093 220075-090
Isopropyl acetate ≥99.6%
Isopropyl acetate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 85 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.8732 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008877
CAS Number: 108-21-4
EINECS: 203-561-1
UN: 1220
ADR: 3,II
Merck Index: 13,05225

Order Now

Specification Test Results

Assay 99.6%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR