Methyl acetoacetate ≥99%

Warning

Synonyms: Methyl 3-oxobutanoate

537365-100G 537365-1KG
220075-932EA 18.09 USD
220075-932 220075-933
Methyl acetoacetate ≥99%
Methyl acetoacetate
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: –39 °C
Density: 1.076 g/cm³ (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00008784
CAS Number: 105-45-3
EINECS: 203-299-8
Merck Index: 12,06090

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR