Ethyl acetoacetate ≥99%

Warning

Synonyms: Acetoacetic acid ethyl ester, EAA

537349-1KG 537349-3KG
220075-930EA 78.51 USD
220075-930 220075-931
Ethyl acetoacetate ≥99%
Ethyl acetoacetate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 180.8 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.0188 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR