6-Fluoro-o-anisaldehyde ≥98%

Synonyms: 2-Fluoro-6-mehoxybenzaldehyde, 2-Fluoro-6-methoxybenzaldehyde, 6-Fluoro-2-anisaldehyde

536733-5G 536733-1G
220076-335EA 1028.77 USD
220076-335 220076-334
6-Fluoro-o-anisaldehyde ≥98%
6-Fluoro-o-anisaldehyde
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD01090998
CAS Number: 146137-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR