3,5-Bis(trifluoromethyl)thiophenol ≥97%

Synonyms: 3,5-Bis(trifluoromethyl)benzenethiol

536083-10ML 536083-1ML
220076-302EA 650.46 USD
220076-302 220076-303
3,5-Bis(trifluoromethyl)thiophenol ≥97%
3,5-Bis(trifluoromethyl)thiophenol
Formula: C₈H₄F₆S
MW: 246.18 g/mol
MDL Number: MFCD00042273
CAS Number: 130783-02-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR