2-Chloro-4-nitropyridine-N-oxide

Synonyms: 2-Chloro-4-nitropyridine N-oxide

534269-5G 534269-25G
220076-201EA 47.38 USD
220076-201 220076-200
2-Chloro-4-nitropyridine-N-oxide
2-Chloro-4-nitropyridine-N-oxide
Formula: C₅H₃ClN₂O₃
MW: 174.54 g/mol
Melting Pt: 158 - 159 °C
MDL Number: MFCD00955617
CAS Number: 14432-16-7
EINECS: 238-404-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR