Xylene (mixture of isomers) for histology

Danger

534056-4X4L 534056-4L 534056-500ML
220076-183EA 131.08 USD
220076-183 220076-182 220076-184
Xylene (mixture of isomers) for histology
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: -34 - 34 °C
Density: 0.86 to 0.88 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=5548e25712f7d73e355ede0a7240c901055e1f61&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR