Pyridine-[D₅]

Danger

532975-10G 532975-25G
220076-015EA 47.38 USD
220076-015 220076-016
Pyridine-[D₅]
Pyridine-[D₅]
Formula: C₅D₅N
Boiling Pt: 114.4 °C (1013 hPa)
Melting Pt: –41 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00044639
CAS Number: 7291-22-7
EINECS: 230-720-2
UN: 1282
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR