Methyl-2-iodobenzoate ≥97%

Synonyms: Methyl 2-iodobenzoate, 2-Iodobenzoic acid methyl ester

532851-250G 532851-50G
220076-004EA 199.23 USD
220076-004 220076-005
Methyl-2-iodobenzoate ≥97%
Methyl-2-iodobenzoate
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 272 °C (1013 hPa)
Density: 1.784 g/cm³ (20 °C)
MDL Number: MFCD00016351
CAS Number: 610-97-9
EINECS: 210-243-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR