1H,1H,2H,2H-Heptadecafluoro-1-decanol ≥97%

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanol, 1H,1H,2H,2H-Perfluoro-1-decanol, 1H,1H,2H,2H-Perfluorodecan-1-ol

532789-25G 532789-5G
220075-491EA 179.85 USD
220075-491 220075-492
1H,1H,2H,2H-Heptadecafluoro-1-decanol ≥97%
1H,1H,2H,2H-Heptadecafluoro-1-decanol
Formula: C₁₀H₅F₁₇O
MW: 464.12 g/mol
MDL Number: MFCD00039544
CAS Number: 678-39-7
EINECS: 211-648-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR