1-Ethyl-3-methylimidazolium chloride

Warning

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride

53096-5G 53096-25G
220075-762EA 66.6 USD
220075-762 220075-333
1-Ethyl-3-methylimidazolium chloride
1-Ethyl-3-methylimidazolium chloride
Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
Melting Pt: ∼80 °C
Density: ~ 1.1 g/cm³ (20 °C)
Flash Pt: 186 °C
MDL Number: MFCD00074843
CAS Number: 65039-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR