2,5-Diaminopyridine

Warning

Synonyms: Pyridine-2,5-diamine, Pyridine-2,5-diyldiamine

530603-5G 530603-10G 530603-1G
220075-751EA 388.77 USD
220075-751 220075-749 220075-750
2,5-Diaminopyridine
2,5-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 329 °C (1013 hPa)
Melting Pt: 110.3 °C
Density: 1.251 g/cm³ (20 °C)
MDL Number: MFCD00129023
CAS Number: 4318-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR