Propagyl bromide ≥80% in xylene

Synonyms: 3-Bromopropyne

530409-125G 530409-50G
220075-726EA 228.31 USD
220075-726 220075-727
Propagyl bromide ≥80% in xylene
Propagyl bromide
Formula: C₃H₃Br
MW: 118.96 g/mol
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR