3-Chloro-2-fluoroaniline ≥97%

530174-25G 530174-5G
220075-710EA 289.69 USD
220075-710 220075-711
3-Chloro-2-fluoroaniline ≥97%
3-Chloro-2-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 99 °C (14 -rr)
MDL Number: MFCD00069415
CAS Number: 2106-04-9
EINECS: 218-283-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR