Magnesium oxide ≥99.995%

Synonyms: Magnesia usta

529699-10G 529699-50G
220075-687EA 212.31 USD
220075-687 220075-688
Magnesium oxide ≥99.995%
Magnesium oxide
Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: 2800 °C
Density: 3.58 g/cm³ (25 °C)
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR