Manganese(IV) oxide

Danger

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

529664-25G 529664-5G
220072-344EA 343.08 USD
220072-344 220075-683
Manganese(IV) oxide
Manganese(IV) oxide
Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C
Density: 5.03 g/cm³ (20 °C)
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR