Iron(III) oxide

Warning

529311-5G 529311-25G
220072-066EA 148.46 USD
220072-066 220072-065
Iron(III) oxide
Iron(III) oxide
Formula: Fe₂O₃
Melting Pt: 1538 °C
Density: 5.24 g/cm³ (20 °C)
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR