2,3,5-Trifluorobenzaldehyde ≥97%

Warning

529192-5ML 529192-1ML
220075-409EA 264.23 USD
220075-409 220075-408
2,3,5-Trifluorobenzaldehyde ≥97%
2,3,5-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 165 °C (1013 hPa)
Density: 1.407 g/cm³ (20 °C)
MDL Number: MFCD00083538
CAS Number: 126202-23-1
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR