3-Iodobenzonitrile ≥99%

529036-10G 529036-1G
220075-123EA 162.62 USD
220075-123 220075-124
3-Iodobenzonitrile ≥99%
3-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
MDL Number: MFCD00079762
CAS Number: 69113-59-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR