2-Bromo-5-fluorobenzaldehyde ≥96%

528978-5G 528978-1G
220075-118EA 101.77 USD
220075-118 220075-117
2-Bromo-5-fluorobenzaldehyde ≥96%
2-Bromo-5-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203.01 g/mol
MDL Number: MFCD00142872
CAS Number: 94569-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR