2-Bromo-4,6-xylidine ≥97%

Warning

Synonyms: 6-Bromo-2,4-dimethylaniline, 6-Bromo-2,4-xylidine, 2-Bromo-4,6-dimethylaniline

528862-1G 528862-10G
220075-395EA 28.22 USD
220075-395 220075-394
2-Bromo-4,6-xylidine ≥97%
2-Bromo-4,6-xylidine
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD00047826
CAS Number: 41825-73-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR