1-Hexanesulfonic acid sodium salt monohydrate ≥98.0% (by titrimetric analysis)

Synonyms: Sodium 1-hexanesulphonate monohydrate

52865-10G 52865-50G 52865-250G
220075-670EA 105.11 USD
220075-670 220075-672 220075-671
1-Hexanesulfonic acid sodium salt monohydrate ≥98.0% (by titrimetric analysis)
1-Hexanesulphonic acid sodium salt monohydrate
Formula: C₆H₁₃NaO₃S·1H₂O
MW: 206.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149549
CAS Number: 207300-91-2
EINECS: 220-601-3

Order Now

Specification Test Results

Assay 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR