5-Fluoro-2-methoxybenzonitrile ≥98%

Synonyms: 2-Cyano-4-fluoroanisole

527734-5G 527734-1G
220075-602EA 98.08 USD
220075-602 220075-601
5-Fluoro-2-methoxybenzonitrile ≥98%
5-Fluoro-2-methoxybenzonitrile
Formula: C₈H₆FNO
MW: 151.14 g/mol
MDL Number: MFCD02683503
CAS Number: 189628-38-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR