1,3-Hexadiene cis- and trans-mixture

Synonyms: Hexa-1,3-diene

527408-1G 527408-5G
220075-063EA 149.69 USD
220075-063 220075-064
1,3-Hexadiene cis- and trans-mixture
1,3-Hexadiene
Formula: C₆H₁₀
MW: 82.15 g/mol
MDL Number: MFCD00009385
CAS Number: 592-48-3
EINECS: 209-759-4
UN: 2458
ADR: 3,II

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR