3-Chloro-2-fluorobenzaldehyde ≥96%

Warning

526606-1G 526606-5G
220075-431EA 28.97 USD
220075-431 220075-432
3-Chloro-2-fluorobenzaldehyde ≥96%
3-Chloro-2-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
MDL Number: MFCD01631571
CAS Number: 85070-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR