4-(Dimethylamino)benzoyl chloride ≥97%

526118-5G 526118-1G
220074-388EA 96.28 USD
220074-388 220074-387
4-(Dimethylamino)benzoyl chloride ≥97%
4-(Dimethylamino)benzoyl chloride
Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
Boiling Pt: 180 °C (15 -rr)
Melting Pt: 147 - 148 °C
MDL Number: MFCD00236039
CAS Number: 4755-50-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR