Ethylene di(acetate) ≥99%

Synonyms: Ethane-1,2-diyl diacetate, Ethylene glycol diacetate, Ethylene Glycol Diacetate, 1,2-Diacetoxyethane

525200-4L 525200-20L 525200-500ML
220075-228EA 187.38 USD
220075-228 220075-227 220075-229
Ethylene di(acetate) ≥99%
Ethylene di(acetate)
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 190…191 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.104 g/cm³ (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00008718
CAS Number: 111-55-7
EINECS: 203-881-1
Merck Index: 13,03833

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR