Triacetin (glycerol triacetate) ≥99%

Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate

525073-1L 525073-4L 525073-20L
220075-199EA 98.65 USD
220075-199 220075-201 220075-200
Triacetin (glycerol triacetate) ≥99%
Triacetin (glycerol triacetate)
Formula: C₉H₁₄O₆
MW: 218.21 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.1596 g/cm³ (20 °C)
Flash Pt: 138 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008716
CAS Number: 102-76-1
EINECS: 203-051-9
Merck Index: 13,09664

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR