4-Aminophthalic acid ≥97%

524716-1G 524716-10G
220075-035EA 136.25 USD
220075-035 220075-034
4-Aminophthalic acid ≥97%
4-Aminophthalic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD00013985
CAS Number: 5434-21-9
EINECS: 226-596-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR