3,4,5-Trifluorophenylboronic acid

Synonyms: (3,4,5-Trifluorophenyl)metaboric acid, Dihydroxy(3,4,5-trifluorophenyl)borane, 3,4,5-Trifluorobenzeneboronic acid, (3,4,5-Trifluorophenyl)boronic acid

524700-1G 524700-5G
220074-207EA 118.46 USD
220074-207 220075-033
3,4,5-Trifluorophenylboronic acid
3,4,5-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
Melting Pt: 292 °C
Storage Temperature: Ambient
MDL Number: MFCD02093069
CAS Number: 143418-49-9
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR