2,4,5-Trifluorophenylboronic acid

524689-25G 524689-5G
220074-204EA 136.25 USD
220074-204 220074-205
2,4,5-Trifluorophenylboronic acid
2,4,5-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
MDL Number: MFCD01863165
CAS Number: 247564-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR