2,4,6-Trifluorophenylboronic acid

Synonyms: (2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane

524107-5G 524107-1G
220074-992EA 128.08 USD
220074-992 220074-991
2,4,6-Trifluorophenylboronic acid
2,4,6-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
MDL Number: MFCD01863169
CAS Number: 182482-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR