4-Bromo-3-nitrobenzaldehyde ≥97%

523151-1G 523151-5G
220074-910EA 57.29 USD
220074-910 220074-911
4-Bromo-3-nitrobenzaldehyde ≥97%
4-Bromo-3-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
MW: 230.02 g/mol
MDL Number: MFCD00204031
CAS Number: 163596-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR