2-Hydroxyphenylboronic acid pinacol ester

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

522554-5G 522554-1G
220074-509EA 162 USD
220074-509 220074-508
2-Hydroxyphenylboronic acid pinacol ester
2-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
MDL Number: MFCD02093754
CAS Number: 269409-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR