2-Amino-6-bromopyridine

Warning

Synonyms: 6-Bromo-2-pyridylamine, 6-Bromopyridin-2-amine

521744-5G 521744-25G
220074-183EA 46.09 USD
220074-183 220074-182
2-Amino-6-bromopyridine
2-Amino-6-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
MDL Number: MFCD00137843
CAS Number: 19798-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR