4-Cyanophenylboronic acid

Warning

Synonyms: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

521418-10G 521418-1G
220074-830EA 240.92 USD
220074-830 220074-831
4-Cyanophenylboronic acid
4-Cyanophenylboronic acid
Formula: C₇H₆BNO₂
MW: 146.94 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD01318968
CAS Number: 126747-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR