2-Cyanophenylboronic acid

Warning

Synonyms: (2-Cyanophenyl)metaboric acid, 2-Cyanobenzeneboronic acid, (2-Cyanophenyl)boronic acid, Dihydroxy(2-cyanophenyl)borane

521396-1G 521396-5G
220074-828EA 38.28 USD
220074-828 220074-829
2-Cyanophenylboronic acid
2-Cyanophenylboronic acid
Formula: C₇H₆BNO₂
MW: 146.94 g/mol
MDL Number: MFCD01632208
CAS Number: 138642-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR