Potassium hexachloroplatinate(IV) ≥99.9%

Danger

520861-1G 520861-5G
220074-783EA 90.69 USD
220074-783 220074-784
Potassium hexachloroplatinate(IV) ≥99.9%
Potassium hexachloroplatinate(IV)
Formula: K₂PtCl₆
Melting Pt: 250 °C
Density: 3.344 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3290
ADR: 6.1,II
Merck Index: 13,07720

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR