Potassium tetrachloroplatinate(II) ≥99.9%

Danger

Synonyms: Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate

520853-1G 520853-25G 520853-5G
220074-780EA 246.15 USD
220074-780 220074-781 220074-782
Potassium tetrachloroplatinate(II) ≥99.9%
Potassium tetrachloroplatinate(II)
Formula: Cl₄K₂Pt
MW: 415.09 g/mol
Boiling Pt: 35 - 40 °C (20 -rr)
Melting Pt: 265 °C
Density: 3.38 g/cm³ (20 °C)
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III
Merck Index: 12,07855

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR