Rhodium(III) chloride hydrate

Synonyms: Rhodium trichloride hydrate

520772-5G 520772-1G
220074-767EA 1758.46 USD
220074-767 220074-766
Rhodium(III) chloride hydrate
Rhodium (III) chloride hydrate
Formula: Cl₃Rh·H₂O
MW: 227.28 g/mol
MDL Number: MFCD00149839
CAS Number: 20765-98-4
EINECS: 233-165-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR