Palladium(II) acetate ≥99.9%

Synonyms: Palladium diacetate, Palladium (II) acetate

520764-5G 520764-25G 520764-1G
220074-765EA 700.31 USD
220074-765 220074-764 220074-763
Palladium(II) acetate ≥99.9%
Palladium(II) acetate
Formula: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR