Palladium(II) chloride

Warning

Synonyms: Palladium dichloride

520659-5G 520659-25G 520659-1G
220074-752EA 468.69 USD
220074-752 220074-751 220074-750
Palladium(II) chloride
Palladium(II) chloride
Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678 °C
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 13,07058

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR