Platinium(IV) oxide hydrate

Synonyms: Platinum(IV) oxide xhydrate, Adam's type catalyst, Platinum dioxide hydrate, Platinum(IV) oxide hydrate, ADAMS' CATALYST

520616-5G 520616-1G
220074-741EA 502.62 USD
220074-741 220074-740
Platinium(IV) oxide hydrate
Platinium(IV) oxide hydrate
Formula: O₂Pt·H₂O
MW: 245.09 g/mol
Melting Pt: 450 °C
Density: 10.2 g/cm³ (20 °C)
MDL Number: MFCD00066964
CAS Number: 52785-06-5
EINECS: 215-223-0
UN: 1479
ADR: 5.1,II
Merck Index: 12,07686

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR