2-Methyl-5-nitroindole

Synonyms: 2-Methyl-5-nitro-1H-indole

520594-5G 520594-1G
220074-737EA 287.69 USD
220074-737 220074-736
2-Methyl-5-nitroindole
2-Methyl-5-nitroindole
Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD00090335
CAS Number: 7570-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR