3-Bromobenzophenone ≥98%

Synonyms: (3-Bromophenyl)(phenyl)methanone, m-Bromobenzophenone

519561-1G 519561-5G
220074-485EA 83.68 USD
220074-485 220074-486
3-Bromobenzophenone ≥98%
3-Bromobenzophenone
Formula: C₁₃H₉BrO
MW: 261.12 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 74.5…77.5 °C
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00672015
CAS Number: 1016-77-9
EINECS: 213-808-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR