3,5-Bis(trifluoromethyl)benzenesulfonyl chloride ≥97%

Synonyms: 3,5-Bis(trifluoromethyl)benzene-1-sulphonyl chloride

519367-5G 519367-1G
220074-480EA 287.69 USD
220074-480 220074-479
3,5-Bis(trifluoromethyl)benzenesulfonyl chloride ≥97%
3,5-Bis(trifluoromethyl)benzenesulphonyl chloride
Formula: C₈H₃ClF₆O₂S
MW: 312.62 g/mol
Melting Pt: 36 - 38 °C
MDL Number: MFCD00014725
CAS Number: 39234-86-1
EINECS: 254-371-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR